Dichloromethane, low water content ≥99.5% stabilised, BakerDRY™, J.T.Baker®
Catalog # 9295-12
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth, septum-sealed
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.5 %
- ACS - Assay (by GC, corrected for water, exclusive of preservative):≥99.5 %
- Appearance:Passes Test
- ACS - Color (APHA):≤10
- ACS - Free Halogens:Passes Test
- ACS - Preservative (cyclohexene), For Information Only:= 2 ppm
- ACS - Residue after Evaporation:≤2 ppm
- ACS - Titrable Acid (µeq/g):≤0.2
- ACS - Water (by KF, coulometric):≤60 ppm
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.5 °C
Melting Pt:–95 °C
Density:1.318 g/cm³
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Stabilised with cyclohexene
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881