Sodium propionate
Catalog # EHERC16493300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:96.06 g/mol
Merck Index:12,08816
Melting Pt:288.76 °C
Flash Pt:157 °C
EINECS:205-290-4
:GHS07
Storage Temperature:Ambient
CAS Number:137-40-6
Formula:C₃H₅NaO₂
MDL Number:MFCD00002759