Cerotic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:506-46-7
:
Formula:C₂₆H₅₂O₂
MDL Number:MFCD00002811
:396.7 g/mol
Boiling Pt:250 °C
Melting Pt:87…89 °C
Density:0.81 g/cm³
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:208-040-2
Specification Test Results
- Appearance:White Glistening powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together