Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Margaric acid ≥98%

Margaric acid ≥98%
Certificates
Margaric acid ≥98%
Margaric acid ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 506-12-7 Molecular Formula: C17H34O2 Formula Weight: 270.46 Melting Point: 59-63° Boiling Point: 227°/100mm Merck Reference: 14,5751 MDL No.: MFCD00002751

Synonyms:  Heptadecanoic acid, Heptadecylic acid
  • CAS Number:506-12-7
  • Formula:C₁₇H₃₄O₂
  • MDL Number:MFCD00002751
  • Molecular Weight:270.46 g/mol
  • Merck Index:14,5751
  • Boiling Pt:227 °C (100 mmHg)
  • Melting Pt:59…63 °C
  • Density:0.853
  • Storage Temperature:Ambient
  • EINECS:208-027-1
Product Family Options

Product Information