Order Entry
Spain
ContactUsLinkComponent
Acetic acid
Acetic acid
  PRLSMC307956.5000
 :  
undefined
Acetic acid
  PRLSMC307956.5000
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190
 :
 :60.05 g/mol
 :118 °C (1013 hPa)
 :17 °C
 :1.05 g/cm³ (20 °C)
 :38.5 °C
 :Ambient
 :200-580-7
 :2789
 :8,II
 :01-2119475328-30
 :GHS02
 :GHS05
 :64-19-7
 :H₃CCOOH
 :MFCD00036152