Arachidic acid 98%
:506-30-9
:
:C₂₀H₄₀O₂
:MFCD00002755
:312.54 g/mol
:12,00802
:328 °C (760 mmHg)
:74…77 °C
:liquid 0,838 g/cm³ (80 °C)
:Ambient
:208-031-3
- Appearance:White to almost white Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:73 to 78 °C
- GC:≥97.5 %
- Titration with TBAH:97.5 to 102.5 %
- Solubility:Passes test
- Sensitivity:(to sulfanilamide) Passes test
Frequently Bought Together