

Acetone ≥99.5%, GPR RECTAPUR®
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:01-2119471330-49
:GHS02
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Related substances:Passes test
- Boiling point:56 - 56.7 °C
- Density (20/4):0.79 - 0.793
- Evaporation residue:Max. 20 ppm
- Assay:Min. 99.5 %
- Acidity:Passes test
- Alkalinity:Passes test
- Appearance of solution:Passes test
- Insolubility in water:Passes test
- IR Spectrum:Passes test
- Substances reducing KMnO4 (as O):Max. 2 ppm
- Water:Max. 0.3 %
1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
More
20065_ACETONE GPR RECTAPUR_BSE_REV1
BSE/TSE Certificates
spec_VWRC20065.293 spec_VWRC20065.327 spec_VWRC20065.362 spec_VWRC20065.464 spec_VWRC20065.470
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together