3-Octanone ≥98%
About this item
Purity: 98% CAS No.: 106-68-3 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 51°(123°F) Refractive Index: 1.4150 Melting Point: -23° Boiling Point: 167-168° Beilstein Reference: 1700021 Merck Reference: 14,6084 UN No.: 1224 MDL No.: MFCD00009515
- CAS Number:106-68-3
- Formula:C₈H₁₆O
- MDL Number:MFCD00009515
- Molecular Weight:128.21 g/mol
- Merck Index:14,6084
- Boiling Pt:167…168 °C
- Melting Pt:–23 °C
- Density:0.822
- Flash Pt:51 °C (123 °F)
- Storage Temperature:Ambient
- EINECS:203-423-0
- UN:1224
- ADR:3,III
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 106-68-3 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 51°(123°F) Refractive Index: 1.4150 Melting Point: -23° Boiling Point: 167-168° Beilstein Reference: 1700021 Merck Reference: 14,6084 UN No.: 1224 MDL No.: MFCD00009515
- CAS Number:106-68-3
- Formula:C₈H₁₆O
- MDL Number:MFCD00009515
- Molecular Weight:128.21 g/mol
- Merck Index:14,6084
- Boiling Pt:167…168 °C
- Melting Pt:–23 °C
- Density:0.822
- Flash Pt:51 °C (123 °F)
- Storage Temperature:Ambient
- EINECS:203-423-0
- UN:1224
- ADR:3,III
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



