Order Entry
Spain
Orders LinkContactUsLinkComponent
Sodium hydroxide 2% in aqueous solution

Sodium hydroxide 2% in aqueous solution

Plastic container, 25 L
Catalog # PRLSMC310002.9025
Corrosive
Supplier:   PROLABO CMP
Sodium hydroxide 2% in aqueous solution
Catalog # PRLSMC310002.9025
Supplier:  PROLABO CMP
CAS Number:  1310-73-2
Sodium hydroxide 2% in aqueous solution

Plastic container, 25 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:13,06608
  • Boiling Pt:120,5 °C (1013 hPa)
  • Melting Pt:-42 °C
  • Density:1,4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-714-2
  • UN:1824
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic container
  • Pk:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.