Digital Solutions
Applications
Support
About
Spain
ContactUsLinkComponent
di-Sodium hydrogen phosphate
Disodium hydrogen phosphate
Disodium hydrogen phosphate
Disodium hydrogen phosphate
RC-095
:
G-Biosciences
:
Disodium hydrogen phosphate
RC-095
:
G-Biosciences
:
Pack type:
Glass bottle for solids
Pk:
500 g
Seq:
3645
:
:
141.96 g/mol
:
12,08805
:
250 °C
:
1.53 g/cm³ (20 °C)
:
Ambient
:
231-448-7
:
01-2119489797-11
:
7558-79-4
:
Na₂HPO₄
:
MFCD00003496