Order Entry
Spain
ContactUsLinkComponent
Dimethyl sulphoxide
Dimethyl sulphoxide
Catalog # HYPETS-20684
CAS Number:  
undefined
Dimethyl sulphoxide
Catalog # HYPETS-20684
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089