
N,N-Dimethylbenzamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:611-74-5
:
Formula:C₉H₁₁NO
MDL Number:MFCD00008320
:149.19 g/mol
Boiling Pt:132…133 °C (15 mmHg)
Melting Pt:42…45 °C
Density:1.11 g/cm³ (23 °C)
Flash Pt:>110 °C (230 °F)
EINECS:210-279-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together