Order Entry
Spain
ContactUsLinkComponent
 
 
Benzamidine hydrochloride hydrate, Sigma-Aldrich®
 :  Merck
 
 
Benzamidine hydrochloride hydrate, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    80 - 84 °C
  • pH:
    5 (50 g/l, H2O, 20 °C)

 

 

Frequently Bought Together