Order Entry
Spain
ContactUsLinkComponent
 
Flammable
(4-Carboxybutyl)triphenylphosphonium bromide, Sigma-Aldrich®
Catalog # 8.18330.0050
Supplier:  Merck
CAS Number:  
Flammable
(4-Carboxybutyl)triphenylphosphonium bromide, Sigma-Aldrich®
Catalog # 8.18330.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    204 - 208 °C
  • Molar Mass:
    443.31 g/mol
  • Bulk Density:
    360 - 400 kg/m3
  • Flash Point:
    195 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

About this item

 :
 :443.32 g/mol
Melting Pt:205…208 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
 :M-Clarity™ quality level = MQ200
EINECS:241-782-5
 :GHS02
CAS Number:17814-85-6
Formula:C₂₃H₂₄BrO₂P
MDL Number:MFCD00011906