


1-Bromobutane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-65-9
:
Formula:BrH₂C(CH₂)₂CH₃
MDL Number:MFCD00000260
:137.02 g/mol
Merck Index:13,01552
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–112 °C
Density:1.276 g/cm³ (20 °C)
EINECS:203-691-9
UN:1126
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:16 °C
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.6 g/l (30 °C)
- Melting Point:-112 °C
- Molar Mass:137.02 g/mol
- Boiling Point:100 - 104 °C
- Vapor Pressure:52 hPa (20 °C)
- Flash Point:18.3 °C
- Refractive Index:1.4394 (20 °C, 589 nm)
- Explosion Limit:2.6 - 6.6 %(V)
- Density:1.27 g/cm3 (20 °C)
- Ignition Point:265 °C
Specification Test Results
Frequently Bought Together