Order Entry
Spain
ContactUsLinkComponent
Lanthanum(III) nitrate hexahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™
Corrosive
Oxidizing
Lanthanum(III) nitrate hexahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™
Certificates
Corrosive
Oxidizing
Lanthanum(III) nitrate hexahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a., Fluka™

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 10 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 10 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 10 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 50 ppm

Specification Test Results

Related Information

Frequently Bought Together