Order Entry
Spain
ContactUsLinkComponent
Sodium perchlorate monohydrate ≥98.0% (by titrimetric analysis), crystallised, Purum p.a.
Irritant
Health
Oxidizing
Sodium perchlorate monohydrate ≥98.0% (by titrimetric analysis), crystallised, Purum p.a.
Certificates
Irritant
Health
Oxidizing
Sodium perchlorate monohydrate ≥98.0% (by titrimetric analysis), crystallised, Purum p.a.

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • pH (5 %, 25 °C):
    5.0 - 8.0
  • Calcium (Ca):
    Max. 50 ppm
  • Cadmium (Cd):
    Max. 50 ppm
  • Cobalt (Co):
    Max. 50 ppm
  • Copper (Cu):
    Max. 50 ppm
  • Iron (Fe):
    Max. 50 ppm
  • Potassium (K):
    Max. 100 ppm
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 50 ppm
  • Zinc (Zn):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 50 ppm
  • Chloride, Chlorate (as Cl):
    Max. 500 ppm

Specification Test Results

Related Information

Frequently Bought Together