Order Entry
Spain
ContactUsLinkComponent
Sodium perchlorate monohydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Oxidizing
Irritant
Sodium perchlorate monohydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Irritant
Sodium perchlorate monohydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    98,0% – 102,0%
  • pH (20 °C; 5%):
    4.5 - 7.0
  • Heavy Metals (as Pb):
    Max. 5 ppm
  • Total N (Nitrogen):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 30 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Fe (Iron):
    Max. 3 ppm
  • K (Potassium):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together