Perchloric acid 0.1 M in acetic acid, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Density:1.06 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Clear colorless Solution
- Water:≤0.2 % (K.F.)
- Strength:0.0998 to 0.1002 M
Specification Test Results
Frequently Bought Together