Order Entry
Spain
ContactUsLinkComponent
 
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Catalog # 1.04817.0250
Supplier:  Merck
CAS Number:  
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Catalog # 1.04817.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.13 g/cm3 (20 °C)
  • pH:
    5.5 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :74.55 g/mol
Density:1.13 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360