(±)-sec-Butylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13952-84-6
:
Formula:C₄H₁₁N
MDL Number:MFCD00008094
:73.14 g/mol
Merck Index:12,01579
Boiling Pt:63 °C (760 mmHg)
Melting Pt:–104 °C
Density:0.72 g/cm³
Flash Pt:–19 °C
EINECS:237-732-7
UN:2734
ADR:8,I
:GHS06
:GHS05
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3920 to 1.3960 (20°C, 589 nm)
- GC:≥98.5 %
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together