

Silver nitrate 5% in aqueous solution
Catalog # PRLSMC306816.0500
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:169.87 g/mol
EINECS:231-853-9
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414