Order Entry
Spain
ContactUsLinkComponent
 
 
 
Hydrofluoric acid 40%, AnalaR NORMAPUR® analytical reagent
 
undefined
 
 
Hydrofluoric acid 40%, AnalaR NORMAPUR® analytical reagent
 

 

  • Assay:
    39.0 - 41.0 %
  • Heavy metals (as Pb):
    Max. 0.5 ppm
  • SiF6 (Hexafluorosilicate):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • PO4 (Phosphate):
    Max. 1 ppm
  • SO4 + SO3 (as SO4):
    Max. 5 ppm
  • Ag (Silver):
    Max. 0.02 ppm
  • Al (Aluminium):
    Max. 0.05 ppm
  • As (Arsenic):
    Max. 0.05 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Be (Beryllium):
    Max. 0.02 ppm
  • Bi (Bismuth):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.2 ppm
  • Ge (Germanium):
    Max. 0.05 ppm
  • Li (Lithium):
    Max. 0.02 ppm
  • Mg (Magnesium):
    Max. 0.2 ppm
  • Mn (Manganese):
    Max. 0.05 ppm
  • Mo (Molybdenum):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Na (Sodium):
    Max. 0.2 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.05 ppm
  • Sr (Strontium):
    Max. 0.02 ppm
  • Ti (Titanium):
    Max. 0.1 ppm
  • Tl (Thallium):
    Max. 0.05 ppm
  • V (Vanadium):
    Max. 0.05 ppm
  • Zn (Zinc):
    Max. 0.05 ppm

 

Frequently Bought Together