

Hydrochloric acid 10% (w/v), EMPROVE® EXPERT Ph. Eur., JP, NF for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (alkalimetric):9.5 - 10.0 %
- Identity:passes test
- Color:≤ 10 Hazen
- Appearance:passes test
- Bromide or iodide:passes test
- Free bromine or chlorine:passes test
- Free chlorine (Cl):≤ 0.0001
- Sulfate (SO₄):≤ 0.0005 %
- Sulphite (SO3):≤ 0.001 %
- Heavy metals (as Pb):≤ 0.0001 %
- As (Arsenic):≤ 0.000015 %
- Hg (Mercury):≤ 0.0000010 %
- Residual solvents (ICH Q3C):...excluded by production process
- Evaporation residue:≤ 0.01
- Residue on ignition (as SO₄):≤ 0.008 %
- Endotoxins:≤ 2.5 I.U./ml
Specification Test Results
Related Information
Frequently Bought Together