3-Methylcyclohexanol for synthesis, Sigma-Aldrich®
Catalog # 8.05807.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:-41 °C
- Molar Mass:114.19 g/mol
- Boiling Point:171 - 173 °C
- Vapor Pressure:1.7 hPa (25 °C)
- Flash Point:70 °C
- Refractive Index:1.4578 (20 °C, 589 nm)
- Explosion Limit:1.3 %(V)
- Density:0.92 g/cm3 (20 °C)
- Ignition Point:295 °C
- Hill Formula:C7H14O
Specification Test Results
About this item
:
:114.19 g/mol
Boiling Pt:171…173 °C (1013 hPa)
Melting Pt:–74 °C
Density:0.9158 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:69 °C
Storage Temperature:Ambient
EINECS:209-709-1
CAS Number:591-23-1
Formula:C₇H₁₄O
MDL Number:MFCD00001446