Thallium ≥99.99% (metals basis), granules ≤ 6 mm (0.2 in)
:7440-28-0
:
:Tl
:MFCD00134063
:204.38 g/mol
:12,09391
:1457 °C (1013 hPa)
:301 °C
:11.85 g/cm³ (20 °C)
:1707
:6.1,II
:Packaged in water.
:231-138-1
:GHS06
:GHS08
:Ambient
- Environmentally Preferable:
Frequently Bought Together