Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Penicillinase (beta-Lactamase)
Penicillinase (beta-Lactamase)
Penicillinase (beta-Lactamase)
DIFC211898
:
DIFCO BD
:
Penicillinase (beta-Lactamase)
DIFC211898
:
DIFCO BD
:
Pack type:
Glass bottle
Pk:
200 mL
CAS No.:
9001-74-5
Storage conditions:
Refrigerator
Enzyme name:
Penicillinase (beta-Lactamase)