Order Entry
Spain
ContactUsLinkComponent
Low viscosity adapter, UL Adapter
Low viscosity adapter, UL Adapter
Catalog # 645-0019
Supplier:  AMETEK
Low viscosity adapter, UL Adapter
Catalog # 645-0019
Supplier:  AMETEK

Specifications

  • Description:
    Enhanced UL adapter for low viscosity liquids
  • Pk:
    1

Specifications

About this item

The UL adapter is designed for measurements on materials with low viscosities in conjunction with LV series. The new spindle with universal connector improves the design and saves significant time on set up and dismantling of equipment.

  • Accurate measurement with range typically 1 to 10 cp at 60 rpm
  • Stainless steel parts are easily cleaned
  • Small sample size of 16 ml
Delivery: Supplied with Locating Channel Assembly, spindle (304 s/s) universal coupling nut, collar assembly with thumbwheel, water jacket, chamber six tube end caps.