Rotational digital rheometer Model DV-III Ultra
Supplier: AMETEK
Certificates
About this item
The ‘all-in-one’ tool to predict a complete flow behaviour.
- Built-in RTD temperature probe
- Bi-directional RS232 provides optional computer control and automatic data gathering capability
- Download custom test programmes with RheoLoader software
- Displayed information includes viscosity, temperature, shear rate/stress, % torque, speed/spindle, step program status and math model calculation
Ordering information: Incl. case, stand, spindle set, temperature probe, EZ-Yield software, RheoLoader software − Cones/plate versions are also available. Please request further details.
Frequently Bought Together