Order Entry
Spain
Orders LinkContactUsLinkComponent
Analogue hygrometer
Analogue hygrometer
Supplier:   Amarell
Analogue hygrometer
Supplier:  Amarell
Certificates
Analogue hygrometer
Analogue hygrometer
2 Options Available Below
Product Details & Documents
This precision wall hygrometer is suitable for indoor applications. Brass and stainless steel case. Mercury filling.
  • Reliable model
  • Accuracy ±5,0%
Product Family Options
Product Information
  • Range
    Division
    Ø
    Availability
    Price
  • 0 - 100 % RH
    1,0 % RH
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 42,10 €
    42,10 €
    1 * 1 UN
       42,10 €
  • 20 - 100 % RH
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 27,80 €
    27,80 €
    1 * 1 UN
       27,80 €