Order Entry
Spain
Orders LinkContactUsLinkComponent
Hotplates
Hotplates
Hotplates
Hotplates
Ceramic top plate or aluminium/silicon alloy top plate.
  • 'Hot' warning light flashes whenever the plate temperature is above 50 °C
Ceramic: excellent chemical resistance, easy to clean and allows higher temperatures.
Aluminium/silicon alloy: efficient and fast rise in temperature.
Product Family Options
  • Heating area
    Temp.
    Heating capacity
    Plate size
    Type
    W×D×H
  • 300×300 mm
    Max. 300 °C
    600 W
    300×300 mm
    HP-200D-L-S
    310×375×125 mm
    1 * 1 UN
    1 * 1 UN - 1 470,00 €
    1 470,00 €
    1 * 1 UN
       1 470,00 €
  • 300×500 mm
    Max. 375 °C
    2250 W
    300×500 mm
    HP-200D-XL-C
    510×375×125 mm
    1 * 1 UN
    1 * 1 UN - 1 260,00 €
    1 260,00 €
    1 * 1 UN
       1 260,00 €
  • 300×300 mm
    Max. 450 °C
    1200 W
    300×300 mm
    HP-200D-L-C
    310×375×105 mm
    1 * 1 UN
    1 * 1 UN - 948,00 €
    948,00 €
    1 * 1 UN
       948,00 €