Order Entry
Spain
Orders LinkContactUsLinkComponent
BD Discardit™ II Syringes
BD Discardit™ II Syringes
BD Discardit™ II Syringes
Supplier:  BD Medical
Certificates
BD Discardit™ II Syringes
BD Discardit™ II Syringes
10 Options Available Below
Product Details & Documents
The BD Discardit™ II two-piece syringe is a single-use medical device intended for injection and aspiration of medical fluids, including both bodily fluids (blood, etc.) and medicines. The lubricant included in the barrel material allows smooth advancement of the plunger, leading to reduced sliding force.
  • Clear and transparent barrel for good visualisation of syringe content
  • Leak-tight
  • Accurate graduation suited to user's needs
  • Robust retaining ring to prevent accidental plunger rod pullout during aspiration
  • Design of finger grip flanges ensures stability and comfort during injection
  • Ergonomically designed plunger for single-handed operation
  • Sterile, single-use
  • Natural rubber latex is not part of the material formulation
Product Family Options
BD Discardit™ II syringes with BD Microlance™ 3 needle
Product Information
  • Capacity
    Sterility
    Availability
    Price
  • 615-0031
    Syringe with needle, Luer slip, concentric tip, blue; 23G needle, 25 mm length
    Syringe with needle, Luer slip, concentric tip, blue; 23G needle, 25 mm length
    2 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 100 UN
    1 * 100 UN - 20,00 €
    20,00 €
    1 * 100 UN
       20,00 €
  • 615-0032
    Syringe with needle, Luer slip, eccentric tip, green; 21G needle, 40 mm length
    Syringe with needle, Luer slip, eccentric tip, green; 21G needle, 40 mm length
    5 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 100 UN
    1 * 100 UN - 25,50 €
    25,50 €
    1 * 100 UN
       25,50 €
  • BDAM300512UK
    Syringe with needle, Luer slip, eccentric tip, green; 21G needle, 40 mm length
    Syringe with needle, Luer slip, eccentric tip, green; 21G needle, 40 mm length
    5 ml
    Sterile
    Discontinued
BD Discardit™ II syringes
Product Information
  • Capacity
    Sterility
    Availability
    Price
  • BDAM300199
    Syringe, Luer slip, concentric tip
    Syringe, Luer slip, concentric tip
    2 ml
    Non sterile
    Log in for availability
    Log in for availability
    1 * 5.700 UN
    1 * 5.700 UN - 586,00 €
    586,00 €
    1 * 5.700 UN
       586,00 €
  • 613-3932
    Syringe, Luer slip, concentric tip
    Syringe, Luer slip, concentric tip
    2 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 100 UN
    1 * 100 UN - 14,40 €
    14,40 €
    1 * 100 UN
       14,40 €
  • BDAM309040
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    5 ml
    Non sterile
    Log in for availability
    Log in for availability
    1 * 3.000 UN
    1 * 3.000 UN - 417,00 €
    417,00 €
    1 * 3.000 UN
       417,00 €
  • 613-3951
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    5 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 100 UN
    1 * 100 UN - 19,20 €
    19,20 €
    1 * 100 UN
       19,20 €
  • BDAM301360
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    10 ml
    Non sterile
    Log in for availability
    Log in for availability
    1 * 1.700 UN
    1 * 1.700 UN - 350,00 €
    350,00 €
    1 * 1.700 UN
       350,00 €
  • 613-3952
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    10 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 100 UN
    1 * 100 UN - 28,00 €
    28,00 €
    1 * 100 UN
       28,00 €
  • 613-3916
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    20 ml
    Sterile
    Log in for availability
    Log in for availability
    1 * 80 UN
    1 * 80 UN - 33,30 €
    33,30 €
    1 * 80 UN
       33,30 €
  • BDAM300296UK
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    20 ml
    Sterile
    Discontinued
  • BDAM309210
    Syringe, Luer slip, eccentric tip
    Syringe, Luer slip, eccentric tip
    20 ml
    Non sterile
    Log in for availability
    Log in for availability
    1 * 950 UN
    1 * 950 UN - 303,00 €
    303,00 €
    1 * 950 UN
       303,00 €