Order Entry
Spain
Orders LinkContactUsLinkComponent
VWR®, Laboratory Stands
Supplier:   VWR Collection
VWR®, Laboratory Stands
Supplier:  VWR Collection
Certificates
VWR®, Laboratory Stands
VWR®, Laboratory Stands
9 Options Available Below
Product Details & Documents
Tripod stands (A): Cast iron tripod stand, painted with hammer finish, steel support rod, nickel plated.
  • Threaded hole
  • Support rod (three lengths available with different diameters)
  • Robust, secure stand
- Threaded hole
- Support rod (three lengths available with different diameters)
- Robust, corrosion resistant
Support stands (B): Cast iron stand base, painted with hammer finish, steel support rod.
- Eccentrically-attached support rod
- Firm, non slip stand
- Available in three different sizes
- Robust, corrosion resistant
Support stands (C): Cast iron stand base, painted with hammer finish, steel support rod, nickel plated.
- Centrally-attached support rod
- Firm, non slip stand
- Available in three different sizes
- Robust, corrosion resistant
Product Family Options
Tripod stands (A)
Product Information
  • Length
    Width
    Arm length
    Availability
    Price
  • 241-0090
    Tripod stand (A) with rod (500 mm length, 8 mm Ø)
    Tripod stand (A) with rod (500 mm length, 8 mm Ø)
    -
    - mm
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 135,00 €
    135,00 €
    1 * 1 UN
       135,00 €
  • 241-0091
    Tripod stand (A) with rod (800 mm length, 12 mm Ø)
    Tripod stand (A) with rod (800 mm length, 12 mm Ø)
    -
    - mm
    145 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 104,00 €
    104,00 €
    1 * 1 UN
       104,00 €
  • 241-0092
    Tripod stand (A) with rod (1000 mm length, 16 mm Ø)
    Tripod stand (A) with rod (1000 mm length, 16 mm Ø)
    -
    - mm
    175 mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 141,00 €
    141,00 €
    1 * 1 UN
       141,00 €
Support stands (B)
Product Information
  • Length
    Width
    Arm length
    Availability
    Price
  • 241-0093
    Support stand (B) with rod (500 mm length, 8 mm Ø)
    Support stand (B) with rod (500 mm length, 8 mm Ø)
    225 mm
    140 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 123,00 €
    123,00 €
    1 * 1 UN
       123,00 €
  • 241-0094
    Support stand (B) with rod (800 mm length, 12 mm Ø)
    Support stand (B) with rod (800 mm length, 12 mm Ø)
    250 mm
    160 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 98,30 €
    98,30 €
    1 * 1 UN
       98,30 €
  • 241-0095
    Support stand (B) with rod (1000 mm length, 16 mm Ø)
    Support stand (B) with rod (1000 mm length, 16 mm Ø)
    270 mm
    180 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 137,00 €
    137,00 €
    1 * 1 UN
       137,00 €
Support stands (C)
Product Information
  • Length
    Width
    Arm length
    Availability
    Price
  • 241-0099
    Support stand (C) with rod (500 mm length, 8 mm Ø)
    Support stand (C) with rod (500 mm length, 8 mm Ø)
    300 mm
    140 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 143,00 €
    143,00 €
    1 * 1 UN
       143,00 €
  • 241-0100
    Support stand (C) with rod (800 mm length, 12 mm Ø)
    Support stand (C) with rod (800 mm length, 12 mm Ø)
    340 mm
    160 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 153,00 €
    153,00 €
    1 * 1 UN
       153,00 €
  • 241-0101
    Support stand (C) with rod (1000 mm length, 16 mm Ø)
    Support stand (C) with rod (1000 mm length, 16 mm Ø)
    380 mm
    180 mm
    - mm
    Log in for availability
    Log in for availability
    1 * 1 UN
    1 * 1 UN - 194,00 €
    194,00 €
    1 * 1 UN
       194,00 €