Order Entry
Spain
ContactUsLinkComponent
Storage plates, 96-well
Storage plates, 96-well
Catalog # 732-4885
   Low Manufacturing Impact
undefined
Storage plates, 96-well
Catalog # 732-4885
   Low Manufacturing Impact

Specifications

  • Description:
    Standard microplate, PP, 370 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    100

Specifications

About this item

PP. Ideal for compound storage, screening and culturing applications.

  • 96 round wells - maximum well volume 370 µl
  • U-shaped well bottom
  • Alphanumeric grid-referencing and industry standard footprint
  • Supporting ribs to aid stacking and improve centrifugation performance

Colour: Natural

Colour: Natural