Plate covers, PS and PP
For prevention of contamination and spillage of samples.
- PS plate covers suitable for all 96-well plates
- PP covers can be used for Thermo-Fast® 384-well plates
- Choice of ridged covers for safe stacking of covered plates or non-ridged covers for use with automated systems
- Catalog No:
- 732-4861
- 732-4865
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Frequently Bought Together