Plates, 384-well, non treated, Nunc™
PS. Designed for DNA libraries and high throughput screening applications. Clear plates suitable for colorimetric assays and sample storage, white plates for luminescence assays, and black plates for fluorescence assays.
- Rounded square well geometry reduces the risk of well-to-well contamination
- Flat bottom wells for optical reading
- Standard footprint compatible with automated handling systems
- Working range: 10 - 100 µl per well
- Clear plates available sterile or non sterile
External dimensions: 128×86 mm
Total well volume: 120 µl
Total well volume: 120 µl
- Catalog No:
- 732-2727
- 736-0192
- 732-2724
- 738-0216
- 738-0164
- 734-2182
- 732-2726
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together