Order Entry
Spain
ContactUsLinkComponent
Membrane filters, PTFE, WTP range, Whatman™
Membrane filters, PTFE, WTP range, Whatman™
Certificates
undefined
Membrane filters, PTFE, WTP range, Whatman™
Specifications
  • Catalog No:
  • 516-1743
  • 516-1741
  • 514-0775
  • 514-0776
  • 516-1742
  • Pore size:
  • 1,00 µm
  • 0,20 µm
  • 0,20 µm
  • 1,00 µm
  • 0,50 µm
  • Thickness:
  • 90 µm
  • 130 µm
  • 130 µm
  • 90 µm
  • 120 µm
  • Airflow rate:
  • 17 l/min/cm² (0,7 bar)
  • 4,5 l/min/cm² (0,7 bar)
  • 4,5 l/min/cm² (0,7 bar)
  • 17 l/min/cm² (0,7 bar)
  • 7,5 l/min/cm² (0,7 bar)
  • Max. temperature:
  • 120 °C
  • 120 °C
  • 120 °C
  • 120 °C
  • 120 °C