VWR® Hichrom Spanners
Supplier: Avantor
Certificates
About this item
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Injector wrench for Shimadzu
Supplier: Restek
VWR® PUREImage, Cleanroom Papers
Supplier: VWR Collection
Dymo labelling cassettes 12mm black on white 1 * 1 items
Supplier: KOEHLER TECHNISCHE PRODUKTEN