Order Entry
Spain
Orders LinkContactUsLinkComponent
Overalls, KLEENGUARD* A45
Overalls, KLEENGUARD* A45
Catalog #
Overalls, KLEENGUARD* A45
Catalog #
Supplier:  
Overalls, KLEENGUARD* A45
Overalls, KLEENGUARD* A45
Product Details & Documents
These hooded overalls with zip fastener are made from anti-static, breathable, extremely durable and silicone-free material.
  • Layered for protection from a very broad range of chemical products (type 6) and to reduce lint
  • Particle protection, retains >99% of fibres larger than 1 µm (type 5)
  • Reinforced seams help protect against tearing
  • Hood is designed to allow a mask to be worn and preserve freedom of movement
  • Colour coded badge for easy identification
They provide excellent protection against particles and splashes, and are ideal for use in critical production areas, especially in the paint, pharmaceutical, chemical and electronics industries.
Colour: White
Certifications: Type 5/6, EN 1149-1, EN 1073-2, Cat. III