Order Entry
Spain
Orders LinkContactUsLinkComponent
Laboratory coats, Tyvek® 500
Laboratory coats, Tyvek® 500
Supplier:   DuPont
Laboratory coats, Tyvek® 500
Supplier:  DuPont
Certificates
Laboratory coats, Tyvek® 500
13 Options Available Below
Product Details & Documents
These garments are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Fabric is ultra-low linting
  • With collar and stitched internal seams
  • Anti-static treatment (EN 1149-5) on both sides
  • PL30 with straight cuffs, PL309 + PL30NP with elasticated cuffs (not tunnelled)
  • Available with zip or press stud closure, with or without pockets
Typical applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Colour: White
Cat. I
Certifications: (EU) 2016/425; partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents).
Packaging: 50 per box, bulk packed.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-0468
  • 113-0467
  • 113-0469
  • 113-0470
  • 113-3882
  • 113-0472
  • 113-0471
  • 113-0473
  • 113-0474
  • 113-0476
  • 113-0475
  • 113-0477
  • 113-0478
  • Model:
  • PL30, 3 pockets
  • PL30, 3 pockets
  • PL30, 3 pockets
  • PL30, 3 pockets
  • PL309, 2 pockets
  • PL309, 2 pockets
  • PL309, 2 pockets
  • PL309, 2 pockets
  • PL309, 2 pockets
  • PL30NP
  • PL30NP
  • PL30NP
  • PL30NP
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Chemical protection|Laboratory
  • Weight:
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
  • 41 g/m²
Specifications
Product Family Options
PL30: Press stud closure, 3 pockets
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    M
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 428,00 €
    428,00 €
    1 * 50 UN
       428,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0468
    Model
    PL30, 3 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    L
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 428,00 €
    428,00 €
    1 * 50 UN
       428,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0467
    Model
    PL30, 3 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 428,00 €
    428,00 €
    1 * 50 UN
       428,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0469
    Model
    PL30, 3 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 428,00 €
    428,00 €
    1 * 50 UN
       428,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0470
    Model
    PL30, 3 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
PL309: Zip closure, 2 pockets, elasticated cuffs
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    S
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 419,00 €
    419,00 €
    1 * 50 UN
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-3882
    Model
    PL309, 2 pockets
    Closure type
    Zip closure
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    M
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 419,00 €
    419,00 €
    1 * 50 UN
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0472
    Model
    PL309, 2 pockets
    Closure type
    Zip closure
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    L
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 419,00 €
    419,00 €
    1 * 50 UN
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0471
    Model
    PL309, 2 pockets
    Closure type
    Zip closure
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 419,00 €
    419,00 €
    1 * 50 UN
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0473
    Model
    PL309, 2 pockets
    Closure type
    Zip closure
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 419,00 €
    419,00 €
    1 * 50 UN
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0474
    Model
    PL309, 2 pockets
    Closure type
    Zip closure
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
PL30NP: Press stud closure, no pockets, elasticated cuffs
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    M
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 383,00 €
    383,00 €
    1 * 50 UN
       383,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0476
    Model
    PL30NP
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    L
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 383,00 €
    383,00 €
    1 * 50 UN
       383,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0475
    Model
    PL30NP
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 383,00 €
    383,00 €
    1 * 50 UN
       383,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0477
    Model
    PL30NP
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 UN
    1 * 50 UN - 383,00 €
    383,00 €
    1 * 50 UN
       383,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0478
    Model
    PL30NP
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory
    Weight
    41 g/m²