Burettes, straight stopcock
Supplier: HIRSCHMANN
Certificates
About this item
DURAN®, borosilicate glass, class AS.
- Calibrated to deliver 'Ex', conformity certified (DE-M marking), with dated batch identification
- Ring marks at major graduations, black graduations
- With glass key
Scale length: 500 mm.
Scale length: 500 mm
Certifications: DIN ISO 385.
Related Information
Certificates
Search by Lot NumberHirschmann brochure EM-Techcolor -volumetric glassware-
Product Literature
Frequently Bought Together