Order Entry
Spain
ContactUsLinkComponent
Carboys, flat, with spigot, Lowboys™, Nalgene®
Carboys, flat, with spigot, Lowboys™, Nalgene®
Catalog # 216-8671
   Low Manufacturing Impact
undefined
Carboys, flat, with spigot, Lowboys™, Nalgene®
Catalog # 216-8671
   Low Manufacturing Impact

Specifications

  • Thread:
    63 mm
  • Capacity:
    8 l
  • W×D×H:
    368×305×165 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Neck I-Ø:
    44 mm
  • Pk:
    1

Specifications

About this item

HDPE, translucent with PP screw cap and spigot.

  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Stackable, max. stacking height: Three containers

This flat carboy easily fits into narrow shelves or refrigerator spaces. Ideal for storing and dispensing reagents, titration solutions and distilled water. With a convenient integral handle for simple, safe transportation.