Human Recombinant FGF-8 (from E. coli)
Fibroblast growth factor 8 (FGF-8) is a critical mitogenic factor that is required for normal development of the eye, ear, brain, and limb. FGF-8 functions broadly to promote cell proliferation, differentiation, and migration. Overexpression of FGF-8 increases tumor growth and angiogenesis. Human and mouse FGF-8 proteins show 100% homology.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:At or below −20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R 3T3 cell proliferation, ED50 ≤150 ng/ml, bioactivity ≥6,7×10³ units/mg
- Gene ID:P55075/P37237
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Androgen-induced growth factor|AIGF|FGF-8b isoform|Heparin-binding growth factor 8|HBGF-8
- Purity:≥95%
- Molecular weight:22,5 kDa
- Sequence:MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together