Human Recombinant TGF-B 1 (from CHO cells)
Catalog # SHBT100-39-20UG
Supplier: Shenandoah Biotechnology
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 µG
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Seq:1154
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤0,5 ng/ml, bioactivity ≥2,0×10⁶ units/mg
- Gene ID:P01137
- Reconstitution instructions:Sterile 10 mM HCl at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:cartilage-inducing factor|Differentiation inhibiting factor
- Purity:≥95%
- Molecular weight:12,8/25,6 kDa
- Sequence:ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS
- Endotoxin level:Low
- Formulation:0,1% trifluoroacetic acid (TFA) and trehalose in a 20:1 trehalose to protein ratio
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
About this item
Thymus and activation regulated chemokine (TARC), also known as CCL17, is a chemokine that is constitutively produced by thymus tissue and activated peripheral blood mononuclear cells (PBMCs), including dendritic cells. TARC signals through the CCR4 receptor to induce chemotaxis of Type 2 T helper (Th2) cells. TARC is important in asthma and allergic diseases, along with bacterial and viral infections.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.