Order Entry
Spain
ContactUsLinkComponent
Human Recombinant BACH1 (from HEK293T cells)
Human Recombinant BACH1 (from HEK293T cells)
Catalog # ORIGTP309275
Supplier:  OriGene
New Product
CAS Number:  
Human Recombinant BACH1 (from HEK293T cells)
Catalog # ORIGTP309275
Supplier:  OriGene
New Product
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293T cells
  • Species:
    Human
  • Storage conditions:
    Store at −80 °C
  • Protein synonyms:
    BACH-1|BTBD24
  • UniProtKB:
    O14867
  • Protein/peptide name:
    BACH1
  • Purity:
    >80% as determined by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    81,8 kDa
  • Sequence:
    MSLSENSVFAYESSVHSTNVLLSLNDQRKKDVLCDVTIFVEGQRFRAHRSVLAACSSYFHSRIVGQADGELNITLPEEVTVKGFEPLIQFAYTAKLILSKENVDEVCKCVEFLSVHNIEESCFQFLKFKFLDSTADQQECPRKKCFSSHCQKTDLKLSLLDQRDLETDEVEEFLENKNVQTPQCKLRRYQGNAKASPPLQDSASQTYESMCLEKDAALALPSLCPKYRKFQKAFGTDRVRTGESSVKDIHASVQPNERSENECLGGVPECRDLQVMLKCDESKLAMEPEETKKDPASQCPTEKSEVTPFPHNSSIDPHGLYSLSLLHTYDQYGDLNFAGMQNTTVLTEKPLSGTDVQEKTFGESQDLPLKSDLGTREDSSVASSDRSSVEREVAEHLAKGFWSDICSTDTPCQMQLSPAVAKDGSEQISQKRSECPWLGIRISESPEPGQRTFTTLSSVNCPFISTLSTEGCSSNLEIGNDDYVSEPQQEPCPYACVISLGDDSETDTEGDSESCSAREQECEVKLPFNAQRIISLSRNDFQSLLKMHKLTPEQLDCIHDIRRRSKNRIAAQRCRKRKLDCIQNLESEIEKLQSEKESLLKERDHILSTLGETKQNLTGLCQKVCKEAALSQEQIQILAKYSAADCPLSFLISEKDKSTPDGELALPSIFSLSDRPPAVLPPCARGNSEPGYARGQESQQMSTATSEQAGPAEQCRQSGGISDFCQQMTDKCTTDE
  • Concentration:
    >0,05 µg/µl as determined by microplate BCA method
  • Formulation:
    25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol

Specifications

About this item