Culture Media in Bottles
Catalog # LIOF495270
Supplier: LIOFILCHEM
New Product
CAS Number:
Specifications
- Pack type:Bottles
- Pk:6
- Description:Buffered NaCl peptone solution pH 7,0 (Perforable cap)
- Environmentally Preferable:
- Closure type:Perforable cap
- Volume:6× 1000 ml
Specifications
About this item
Buffered NaCl Peptone Solution pH 7.0 is used for dissolving, suspending and diluting test samples.
- Its composition conforms to recommendations in the Harmonized Pharmacopoeias for the microbiological examination of non sterile products
- This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics
- Shelf life two years
The powder is very hygroscopic, store the powder at 10 to 30 °C, in a dry environment, in its original container tightly closed. Store bottles and bags at 10 to 25 °C away from light. Do not use the product beyond its expiry date on the label or if product shows any evidence of contamination or any sign of deterioration.