Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
Ferrule for Tubing
Ferrule for Tubing
Ferrule for Tubing
Catalog # METB51191202
Supplier:
Mettler - Toledo
New Product
Expand image
Ferrule for Tubing
Catalog # METB51191202
Supplier:
Mettler - Toledo
New Product
Specifications
Description:
Ferrule for tubing and sensors
Material:
ETFE
Pk:
4
For tubing ext.Ø:
1,6 mm
Specifications
About this item
Product Details
Product Details