Order Entry
Spain
ContactUsLinkComponent
Acrylamide-d3 (1,0 mg/ml in Deionised Water with 0,1% Formic Acid)
Acrylamide-d3 (1,0 mg/ml in Deionised Water with 0,1% Formic Acid)
Catalog # PROOTRCKIT97922X1M
New Product
CAS Number:  
Acrylamide-d3 (1,0 mg/ml in Deionised Water with 0,1% Formic Acid)
Catalog # PROOTRCKIT97922X1M
New Product
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Description:
    Acrylamide-d3 (1,0 mg/ml in deionised water with 0,1% formic acid)
  • GHS signal:
    Danger
  • Formula:
    C₃H₂D₃NO
  • Molecular weight:
    74,1
  • Storage temperature:
    4 °C

Specifications

About this item