Order Entry
Spain
ContactUsLinkComponent
 
Anti-MATH2/NEUROD6 Rabbit Polyclonal Antibody
  NOVUNBP2298650.025
New Product
Anti-MATH2/NEUROD6 Rabbit Polyclonal Antibody
  NOVUNBP2298650.025
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    MATH2/NEUROD6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    63974
  • Antigen synonyms:
    Math-2|bHLHa2MATH2|BHLHA2|NEX1M|Protein atonal homolog 2|NeuroD6|neurogenic differentiation 6|neurogenic differentiation factor 6|Class A basic helix-loop-helix protein 2|Atoh2
  • Storage buffer:
    50% saturated ammonium sulfate and PBS
  • Storage temperature:
    Store at −70 °C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide
  • Tested applications:
    Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
  • Purification:
    Affinity purified
  • Pk:
    0,25 mg

 

 

 : This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen: MATH2/NEUROD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse