Order Entry
Spain
ContactUsLinkComponent
Benzoic acid ≥99.0%, zone refined: number of passes 20
Irritant
Benzoic acid ≥99.0%, zone refined: number of passes 20
Catalog # TCIAB0062-1SAMPLE
Supplier:  TCI
CAS Number:  
Irritant
Benzoic acid ≥99.0%, zone refined: number of passes 20
Catalog # TCIAB0062-1SAMPLE
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0014

Specifications

Specification Test Results

  • Purity/Analysis Method:
    >99.0% (GC)

Specification Test Results

About this item

 :
 :122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C
Melting Pt:125 °C
Density:1.316 g/cm³ (20 °C)
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
 :GHS07
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398